Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
epg-7
Entrez Gene ID
180996
Gene Name
ATG11 domain-containing protein
Gene Aliases
CELE_T08A9.1
Chromosome Location
Chr.X: 7334714 - 7340268 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076840.8 NP_509241.2 2 - 3 192 114

View Full Product Details