Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
coh-1
Entrez Gene ID
181003
Gene Name
protein-coding
Gene Aliases
CELE_K08A8.3
Chromosome Location
Chr.X: 7465193 - 7475019 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076861.7 NP_509262.2 5 - 6 1178 93

View Full Product Details