Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcca-1
Entrez Gene ID
181026
Gene Name
Propionyl-CoA carboxylase alpha chain, mitochondrial
Gene Aliases
CELE_F27D9.5
Chromosome Location
Chr.X: 7657512 - 7687738 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076892.7 NP_509293.1 2 - 3 118 62

View Full Product Details