Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-18
Entrez Gene ID
181030
Gene Name
Putative acetylcholine regulator unc-18
Gene Aliases
CELE_F27D9.1
Chromosome Location
Chr.X: 7682896 - 7686200 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029435.5 NP_001024606.1 3 - 4 288 127
GenBank mRNA S66176.1 - 3 - 4 252 127

View Full Product Details