Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-12
Entrez Gene ID
187229
Gene Name
Histone-lysine N-methyltransferase
Gene Aliases
CELE_K09F5.5
Chromosome Location
Chr.X: 7733020 - 7736271 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076905.5 NP_509306.2 3 - 4 782 74

View Full Product Details