Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
deg-1
Entrez Gene ID
181035
Gene Name
protein-coding
Gene Aliases
CELE_C47C12.6
Chromosome Location
Chr.X: 7753070 - 7774817 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373369.2 NP_001360711.1 7 - 8 763 66
NM_001373370.2 NP_001360706.1 5 - 5 823 66
NM_001373371.2 NP_001360707.1 5 - 5 823 66
NM_001373372.2 NP_001360708.1 5 - 5 823 66
NM_001373373.2 NP_001360709.1 5 - 5 823 66
NM_001373374.2 NP_001360710.1 5 - 5 823 66
NM_076910.5 NP_509311.1 5 - 6 833 66
GenBank mRNA L34414.1 - 5 - 6 855 66

View Full Product Details