Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sek-1
Entrez Gene ID
181043
Gene Name
Dual specificity mitogen-activated protein kinase kinase sek-1
Gene Aliases
CELE_R03G5.2
Chromosome Location
Chr.X: 7816734 - 7821045 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076921.7 NP_509322.2 5 - 6 940 88
GenBank mRNA AB024087.1 - 5 - 6 950 88
AB060731.2 - 5 - 6 950 88

View Full Product Details