Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tnt-3
Entrez Gene ID
181052
Gene Name
protein-coding
Gene Aliases
CELE_C14F5.3
Chromosome Location
Chr.X: 7937633 - 7945099 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373383.1 NP_001359847.1 8 - 9 3600 68

View Full Product Details