Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-3
Entrez Gene ID
181094
Gene Name
Sulfate permease family protein 3
Gene Aliases
CELE_F41D9.5
Chromosome Location
Chr.X: 8403539 - 8408367 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077023.5 NP_509424.2 4 - 5 587 152
GenBank mRNA AY887909.1 - 4 - 5 587 152

View Full Product Details