Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-10
Entrez Gene ID
181101
Gene Name
Degenerin mec-10
Gene Aliases
CELE_F16F9.5
Chromosome Location
Chr.X: 8465760 - 8470314 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077037.7 NP_509438.1 16 - 17 2065 101
GenBank mRNA L25312.1 - 16 - 17 2007 101

View Full Product Details