Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-18
Entrez Gene ID
181139
Gene Name
TWiK family of potassium channels protein 18
Gene Aliases
CELE_C24A3.6
Chromosome Location
Chr.X: 8988137 - 8991647 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383619.1 6 - 7 974 144
GenBank mRNA AF083650.1 - 6 - 7 926 144

View Full Product Details