Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- tmbi-4
- Entrez Gene ID
-
181150
- Gene Name
- Transmembrane BAX inhibitor motif-containing protein 4
- Gene Aliases
-
CELE_B0563.4
- Chromosome Location
-
Chr.X: 9150694 - 9160660 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077142.8 |
NP_509543.2 |
1 - 2 |
56 |
66 |
View Full Product Details