Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- lgc-4
- Entrez Gene ID
-
181155
- Gene Name
- Ligand-gated ion channel 4
- Gene Aliases
-
CELE_F18G5.4
- Chromosome Location
-
Chr.X: 9235103 - 9242223 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077155.5 |
NP_509556.4 |
3 - 4 |
785 |
109 |
- Gene Symbol
- col-175
- Entrez Gene ID
-
24104282
- Gene Name
- Col_cuticle_N domain-containing protein
- Gene Aliases
-
CELE_C35B8.1
- Chromosome Location
-
Chr.X: 9233638 - 9240564 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383621.1 |
|
1 - 2 |
151 |
109 |
View Full Product Details