Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tba-8
Entrez Gene ID
191455
Gene Name
Tubulin alpha-8 chain
Gene Aliases
CELE_ZK899.4
Chromosome Location
Chr.X: 9455485 - 9458348 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077190.5 NP_509591.2 9 - 10 1258 62

View Full Product Details