Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gap-2
Entrez Gene ID
181172
Gene Name
protein-coding
Gene Aliases
CELE_ZK899.8
Chromosome Location
Chr.X: 9472467 - 9516454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129726.4 NP_001123198.1 6 - 7 1770 94
NM_001307914.4 NP_001294843.1 10 - 11 2236 94
NM_001307916.4 NP_001294845.1 15 - 16 3013 94
NM_001383623.1 13 - 14 2617 94
NM_077193.6 NP_509594.3 15 - 16 3014 94
NM_077194.7 NP_509595.1 16 - 17 3076 94
NM_077195.7 NP_509596.1 14 - 15 3664 94
NM_077196.6 NP_509597.1 13 - 14 2803 94
NM_077197.6 NP_509598.1 12 - 13 2553 94
NM_077198.7 NP_509599.1 10 - 11 3133 94
NM_171753.7 NP_741873.1 13 - 14 3036 94
NM_171754.6 NP_741874.1 6 - 7 1759 94

View Full Product Details