Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gap-2
Entrez Gene ID
181172
Gene Name
protein-coding
Gene Aliases
CELE_ZK899.8
Chromosome Location
Chr.X: 9472467 - 9516454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129726.4 NP_001123198.1 7 - 8 1967 67
NM_001307914.4 NP_001294843.1 11 - 12 2433 67
NM_001383623.1 14 - 15 2814 67
NM_077193.6 NP_509594.3 16 - 17 3211 67
NM_077194.7 NP_509595.1 17 - 18 3273 67
NM_077195.7 NP_509596.1 15 - 16 3861 67
NM_077196.6 NP_509597.1 14 - 15 3000 67
NM_077197.6 NP_509598.1 13 - 14 2750 67
NM_077198.7 NP_509599.1 11 - 12 3330 67
NM_171753.7 NP_741873.1 14 - 15 3233 67
NM_171754.6 NP_741874.1 7 - 8 1956 67

View Full Product Details