Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gap-2
Entrez Gene ID
181172
Gene Name
protein-coding
Gene Aliases
CELE_ZK899.8
Chromosome Location
Chr.X: 9472467 - 9516454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129726.4 NP_001123198.1 9 - 10 2288 90
NM_001307914.4 NP_001294843.1 13 - 14 2754 90
NM_001307916.4 NP_001294845.1 18 - 19 3525 90
NM_001383623.1 16 - 17 3135 90
NM_077193.6 NP_509594.3 18 - 19 3532 90
NM_077194.7 NP_509595.1 19 - 20 3594 90
NM_077195.7 NP_509596.1 17 - 18 4182 90
NM_077196.6 NP_509597.1 16 - 17 3321 90
NM_077197.6 NP_509598.1 15 - 16 3071 90
NM_077198.7 NP_509599.1 13 - 14 3651 90
NM_171753.7 NP_741873.1 16 - 17 3554 90
NM_171754.6 NP_741874.1 9 - 10 2277 90

View Full Product Details