Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gap-2
Entrez Gene ID
181172
Gene Name
protein-coding
Gene Aliases
CELE_ZK899.8
Chromosome Location
Chr.X: 9472467 - 9516454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383623.1 2 - 3 286 92
NM_077194.7 NP_509595.1 5 - 6 745 92
NM_077195.7 NP_509596.1 3 - 4 1333 92
NM_077196.6 NP_509597.1 2 - 3 472 92
NM_171753.7 NP_741873.1 2 - 3 705 92

View Full Product Details