Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gap-2
Entrez Gene ID
181172
Gene Name
protein-coding
Gene Aliases
CELE_ZK899.8
Chromosome Location
Chr.X: 9472467 - 9516454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307914.4 NP_001294843.1 3 - 4 358 86
NM_001307916.4 NP_001294845.1 8 - 9 1135 86
NM_001383623.1 6 - 7 739 86
NM_077193.6 NP_509594.3 8 - 9 1136 86
NM_077194.7 NP_509595.1 9 - 10 1198 86
NM_077195.7 NP_509596.1 7 - 8 1786 86
NM_077196.6 NP_509597.1 6 - 7 925 86
NM_077197.6 NP_509598.1 5 - 6 675 86
NM_077198.7 NP_509599.1 3 - 4 1255 86
NM_171753.7 NP_741873.1 6 - 7 1158 86

View Full Product Details