Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nuc-1
Entrez Gene ID
181174
Gene Name
Deoxyribonuclease-2
Gene Aliases
CELE_C07B5.5
Chromosome Location
Chr.X: 9539611 - 9542329 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077203.8 NP_509604.1 3 - 4 308 98

View Full Product Details