Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-4
Entrez Gene ID
181187
Gene Name
protein-coding
Gene Aliases
CELE_F15G9.4
Chromosome Location
Chr.X: 9717565 - 9753764 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029411.3 NP_001024582.1 13 - 14 4515 115
NM_001373449.3 NP_001359724.1 13 - 13 4454 115
NM_001373450.2 NP_001360633.1 12 - 13 4160 115
NM_001373451.2 NP_001360634.1 12 - 13 4160 115
NM_077234.7 NP_509635.2 13 - 14 4454 115
GenBank mRNA AF074901.1 - 13 - 14 4454 115

View Full Product Details