Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-23
Entrez Gene ID
181205
Gene Name
protein-coding
Gene Aliases
CELE_C17G1.3
Chromosome Location
Chr.X: 9922862 - 9925173 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083286.4 NP_001076755.1 4 - 5 1043 121
NM_001083287.6 NP_001076756.1 3 - 4 673 121

View Full Product Details