Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pssy-1
Entrez Gene ID
181211
Gene Name
PhosphatidylSerine SYnthase
Gene Aliases
CELE_ZC506.3
Chromosome Location
Chr.X: 9965599 - 9968806 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077272.6 NP_509673.1 5 - 6 750 91

View Full Product Details