Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grk-1
Entrez Gene ID
181212
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
CELE_F19C6.1
Chromosome Location
Chr.X: 9991248 - 9999459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077275.8 NP_509676.1 9 - 10 1717 87

View Full Product Details