Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mkk-4
Entrez Gene ID
185682
Gene Name
MAP kinase kinase mkk-4
Gene Aliases
CELE_F42G10.2
Chromosome Location
Chr.X: 10027856 - 10030130 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077281.7 NP_509682.1 7 - 8 849 132

View Full Product Details