Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
moc-1
Entrez Gene ID
181225
Gene Name
Molybdopterin molybdenumtransferase
Gene Aliases
CELE_T06H11.4
Chromosome Location
Chr.X: 10136129 - 10141995 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077299.6 NP_509700.2 6 - 7 1580 69

View Full Product Details