Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sex-1
Entrez Gene ID
181231
Gene Name
Steroid hormone receptor family member cnr14
Gene Aliases
CELE_F44A6.2
Chromosome Location
Chr.X: 10201329 - 10205421 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029491.5 NP_001024662.1 4 - 5 663 91
GenBank mRNA U13074.1 - 4 - 5 684 91

View Full Product Details