Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rme-4
Entrez Gene ID
185870
Gene Name
protein-coding
Gene Aliases
CELE_F46F6.1
Chromosome Location
Chr.X: 10373232 - 10379845 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373469.3 NP_001359519.1 5 - 5 628 127
NM_077338.5 NP_509739.1 6 - 7 819 127

View Full Product Details