Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyf-6
Entrez Gene ID
3565899
Gene Name
protein-coding
Gene Aliases
CELE_F46F6.4
Chromosome Location
Chr.X: 10379693 - 10386508 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083298.2 NP_001076767.1 2 - 3 386 64
NM_001083299.3 NP_001076768.1 2 - 3 386 64
NM_001381083.2 NP_001367144.1 2 - 3 322 64

View Full Product Details