Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-15
Entrez Gene ID
181248
Gene Name
protein-coding
Gene Aliases
CELE_ZC504.4
Chromosome Location
Chr.X: 10421653 - 10428657 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029800.3 NP_001024971.1 5 - 6 874 91
NM_001029801.5 NP_001024972.1 5 - 6 874 91
NM_001029802.5 NP_001024973.1 5 - 6 798 91
NM_001029803.6 NP_001024974.1 5 - 6 881 91
GenBank mRNA AF087131.1 - 5 - 6 896 91

View Full Product Details