Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vab-3
Entrez Gene ID
181251
Gene Name
protein-coding
Gene Aliases
CELE_F14F3.1
Chromosome Location
Chr.X: 10499890 - 10519580 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029399.4 NP_001024570.1 3 - 4 458 78
GenBank mRNA U31537.1 - 3 - 4 412 78

View Full Product Details