Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abl-1
Entrez Gene ID
181261
Gene Name
protein-coding
Gene Aliases
CELE_M79.1
Chromosome Location
Chr.X: 10605827 - 10624207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381086.2 NP_001368776.1 7 - 8 1157 76
NM_001381087.2 NP_001368775.1 9 - 10 1247 76
NM_077376.10 NP_509777.2 8 - 9 1218 76
NM_077377.6 NP_509778.2 9 - 10 1238 76
NM_077378.5 NP_509779.2 5 - 6 1295 76

View Full Product Details