Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clc-4
Entrez Gene ID
188095
Gene Name
CLaudin-like in Caenorhabditis
Gene Aliases
CELE_T05A10.2
Chromosome Location
Chr.X: 10782927 - 10783922 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077399.2 NP_509800.1 2 - 3 225 82

View Full Product Details