Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dgn-1
Entrez Gene ID
181283
Gene Name
Peptidase S72 domain-containing protein
Gene Aliases
CELE_T21B6.1
Chromosome Location
Chr.X: 10920515 - 10928998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381093.1 NP_001366742.1 4 - 5 870 69

View Full Product Details