Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ver-4
Entrez Gene ID
186632
Gene Name
Tyrosine-protein kinase receptor ver-4
Gene Aliases
CELE_F59F3.5
Chromosome Location
Chr.X: 10982381 - 10988267 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077434.4 NP_509835.2 7 - 8 2518 126

View Full Product Details