Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ver-3
Entrez Gene ID
181289
Gene Name
Tyrosine-protein kinase receptor ver-3
Gene Aliases
CELE_F59F3.1
Chromosome Location
Chr.X: 10992477 - 10997846 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077435.6 NP_509836.1 10 - 11 3127 64

View Full Product Details