Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-31
Entrez Gene ID
186484
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_F58A3.5
Chromosome Location
Chr.X: 11008268 - 11009054 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077438.7 NP_509839.1 2 - 3 309 135

View Full Product Details