Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-15
Entrez Gene ID
181291
Gene Name
protein-coding
Gene Aliases
CELE_F58A3.2
Chromosome Location
Chr.X: 11016400 - 11023421 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313426.3 NP_001300355.1 13 - 14 2521 70
NM_001313427.4 NP_001300356.1 13 - 14 2521 70
NM_001313435.5 NP_001300364.1 14 - 14 2357 70
NM_001373496.2 NP_001360406.1 13 - 14 2520 70
NM_001381095.1 NP_001368580.1 13 - 14 2324 70
NM_001381096.1 NP_001368579.1 13 - 14 2324 70
NM_001381098.1 NP_001367004.1 13 - 14 2324 70
NM_001383640.1 13 - 14 2521 70
GenBank mRNA U39761.1 - 13 - 14 2489 70

View Full Product Details