Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apt-9
Entrez Gene ID
189194
Gene Name
AdaPTin or adaptin-related protein
Gene Aliases
CELE_W04G3.4
Chromosome Location
Chr.X: 11068527 - 11072047 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077452.6 NP_509853.2 6 - 7 1000 123

View Full Product Details