Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-7
Entrez Gene ID
189195
Gene Name
protein-coding
Gene Aliases
CELE_W04G3.6
Chromosome Location
Chr.X: 11080678 - 11087474 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038481.4 NP_001033570.1 2 - 3 247 141
NM_001038482.4 NP_001033571.1 2 - 3 247 141
GenBank mRNA AY887915.1 - 2 - 3 269 141
AY887916.1 - 2 - 3 269 141

View Full Product Details