Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hizr-1
Entrez Gene ID
191322
Gene Name
High zinc activated nuclear receptor protein
Gene Aliases
CELE_ZK455.6
Chromosome Location
Chr.X: 11318278 - 11321123 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077492.6 NP_509893.3 1 - 2 187 75

View Full Product Details