Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asm-2
Entrez Gene ID
181323
Gene Name
Sphingomyelin phosphodiesterase 2
Gene Aliases
CELE_ZK455.4
Chromosome Location
Chr.X: 11321076 - 11324251 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077493.7 NP_509894.2 3 - 4 694 82

View Full Product Details