Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aco-1
Entrez Gene ID
181324
Gene Name
Probable cytoplasmic aconitate hydratase
Gene Aliases
CELE_ZK455.1
Chromosome Location
Chr.X: 11339874 - 11343327 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077497.4 NP_509898.1 2 - 3 264 68

View Full Product Details