Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tni-1
Entrez Gene ID
181329
Gene Name
Troponin I 1
Gene Aliases
CELE_F42E11.4
Chromosome Location
Chr.X: 11375878 - 11378717 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077505.7 NP_509906.1 5 - 6 545 127

View Full Product Details