Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slo-2
Entrez Gene ID
181332
Gene Name
protein-coding
Gene Aliases
CELE_F08B12.3
Chromosome Location
Chr.X: 11394605 - 11405931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029357.5 NP_001024528.1 16 - 17 3400 145
NM_001029358.5 NP_001024529.1 16 - 17 3081 145
NM_001270215.3 NP_001257144.1 18 - 19 3192 145
NM_001270216.3 NP_001257145.1 17 - 18 3147 145
NM_001270217.3 NP_001257146.1 17 - 18 3201 145
NM_001270218.3 NP_001257147.1 17 - 18 3162 145
GenBank mRNA AF232770.1 - 16 - 17 3093 145

View Full Product Details