Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-14
Entrez Gene ID
181337
Gene Name
protein-coding
Gene Aliases
CELE_T25C12.1
Chromosome Location
Chr.X: 11465557 - 11485871 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077515.4 NP_509916.2 8 - 9 1102 120
NM_077516.6 NP_509917.1 6 - 7 1195 120

View Full Product Details