Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-2
Entrez Gene ID
181344
Gene Name
protein-coding
Gene Aliases
CELE_C35C5.4
Chromosome Location
Chr.X: 11548608 - 11550740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313405.4 NP_001300334.1 1 - 2 130 69
NM_077530.4 NP_509931.1 3 - 4 517 69

View Full Product Details