Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lev-8
Entrez Gene ID
191600
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C35C5.5
Chromosome Location
Chr.X: 11560915 - 11563707 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077531.7 NP_509932.2 11 - 12 1410 67

View Full Product Details