Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-44
Entrez Gene ID
181349
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_Y71H9A.1
Chromosome Location
Chr.X: 11622967 - 11627113 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077541.4 NP_509942.4 9 - 10 1058 92

View Full Product Details