Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnd-1
Entrez Gene ID
183457
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_C44C10.8
Chromosome Location
Chr.X: 11684561 - 11685788 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077551.4 NP_509952.1 2 - 3 421 81

View Full Product Details